Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001135924.1 | NP_001129396.1 | 18, 19 | 4251 | |
XM_005249641.1 | XP_005249698.1 | 15, 16 | 3365 | ||
XM_011515175.1 | XP_011513477.1 | 18, 19 | 3928 | ||
XM_017011828.1 | XP_016867317.1 | 16, 17 | 3583 | ||
XM_017011829.1 | XP_016867318.1 | 15, 16 | 3252 | ||
GenBank mRNA | AK027618.1 | - | 7, 8 | 1838 | |
BC144319.1 | - | 15, 16 | 3403 | 9052839 |