Gene Transcripts

Gene Symbol
HAUS5
Entrez Gene ID
23354
Gene Name
HAUS augmin like complex subunit 5
Gene Aliases
KIAA0841, dgt5
Chromosome Location
Chr. 19: 35612744 - 35625349 on Build GRCh38
UniGene ID
Hs.7426
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015302.1 NP_056117.1 19 3842
XM_011526684.2 XP_011524986.1 18 3729
GenBank mRNA AB020648.1 - 19 3816

View Full Product Details