Gene Transcripts

Gene Symbol
LEUTX
Entrez Gene ID
342900
Gene Name
leucine twenty homeobox
Gene Aliases
Chromosome Location
Chr. 19: 39776594 - 39786135 on Build GRCh38
UniGene ID
Hs.451638
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001143832.1 NP_001137304.1 2 147
GenBank mRNA CR746510.1 - 2 146

View Full Product Details