Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001080456.2 | NP_001073925.2 | 4 | 1082 | |
XM_005258864.3 | XP_005258921.2 | 5 | 1241 | ||
XM_006723189.3 | XP_006723252.1 | 5 | 1262 | ||
GenBank mRNA | BX094413.1 | - | 3 | 640 |