Gene Transcripts

Gene Symbol
ZNF699
Entrez Gene ID
374879
Gene Name
zinc finger protein 699
Gene Aliases
hang
Chromosome Location
Chr. 19: 9291140 - 9309714 on Build GRCh38
UniGene ID
Hs.633842
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198535.2 NP_940937.1 4, 5 664
GenBank mRNA AK095463.1 - 2, 3 176
BC109267.1 - 3, 4 294 40025983
BC109268.1 - 3, 4 294 40025986

View Full Product Details