Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001142807.1 | NP_001136279.1 | 8 | 834 | |
XM_011511404.2 | XP_011509706.1 | 8 | 868 | ||
XM_011511405.2 | XP_011509707.1 | 8 | 886 | ||
XM_011511406.2 | XP_011509708.1 | 8 | 859 | ||
XM_011511407.2 | XP_011509709.1 | 7 | 619 | ||
XM_011511409.1 | XP_011509711.1 | 8 | 868 | ||
XM_011511411.2 | XP_011509713.1 | 8 | 868 | ||
XM_011511413.1 | XP_011509715.1 | 8 | 868 | ||
XM_011511414.2 | XP_011509716.1 | 8 | 868 | ||
XM_011511415.2 | XP_011509717.1 | 8 | 868 | ||
XM_011511416.2 | XP_011509718.1 | 8 | 868 | ||
XM_011511419.2 | XP_011509721.1 | 8 | 868 | ||
XM_011511420.2 | XP_011509722.1 | 8 | 868 | ||
XM_011511421.2 | XP_011509723.1 | 8 | 868 | ||
XM_011511422.1 | XP_011509724.1 | 8 | 868 | ||
XM_011511423.1 | XP_011509725.1 | 8 | 868 | ||
XM_011511427.2 | XP_011509729.1 | 8 | 868 | ||
XM_011511428.2 | XP_011509730.1 | 8 | 868 | ||
XM_011511429.2 | XP_011509731.1 | 8 | 868 | ||
XM_011511432.2 | XP_011509734.1 | 8 | 696 | ||
XM_017004431.1 | XP_016859920.1 | 8 | 868 | ||
XM_017004432.1 | XP_016859921.1 | 8 | 868 | ||
GenBank mRNA | AK001904.1 | - | 2 | 127 | |
AK300513.1 | - | 8 | 833 | ||
BC022268.1 | - | 8 | 773 | ||
BC131514.1 | - | 2 | 124 | ||
CX788840.1 | - | 8 | 775 |