Gene Transcripts

Gene Symbol
CASC1
Entrez Gene ID
55259
Gene Name
cancer susceptibility candidate 1
Gene Aliases
LAS1, PPP1R54
Chromosome Location
Chr. 12: 25107057 - 25195177 on Build GRCh38
UniGene ID
Hs.407771
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001082972.2 NP_001076441.1 9 1233
NM_001082973.2 NP_001076442.1 8 1025
NM_001204101.2 NP_001191030.1 6 848
NM_001204102.2 NP_001191031.1 10 1081
NM_001319977.1 NP_001306906.1 7 933
NM_001319978.1 NP_001306907.1 8 1199
NM_018272.4 NP_060742.3 9 1043
XM_005253406.1 XP_005253463.1 9 1157
XM_005253408.1 XP_005253465.1 8 919
XM_005253409.1 XP_005253466.1 7 933
XM_006719101.1 XP_006719164.1 9 1157
XM_011520723.1 XP_011519025.1 9 1157
XM_011520726.1 XP_011519028.1 9 1157
XM_011520727.1 XP_011519029.1 9 1157
XM_017019560.1 XP_016875049.1 8 1122
XM_017019561.1 XP_016875050.1 7 1129
XM_017019562.1 XP_016875051.1 8 1007
XM_017019563.1 XP_016875052.1 9 1039
XM_017019564.1 XP_016875053.1 8 1007
XM_017019565.1 XP_016875054.1 9 1157
XM_017019566.1 XP_016875055.1 9 1368
XM_017019567.1 XP_016875056.1 6 812
XM_017019568.1 XP_016875057.1 7 951
GenBank mRNA AK001783.1 - 3 448
AK093102.1 - 9 1011
AK297998.1 - 6 812
AK301763.1 - 7 594
AK302235.1 - 8 989
AK304602.1 - 7 933
AY423543.1 - 7 941
BC028415.1 - 9 1174 4823620
BC031047.1 - 9 1024
BC047415.1 - 10 1079 5269578
BC057795.1 - 8 1023 5266518
BC117281.1 - 6 858
BC117283.1 - 6 858

View Full Product Details