Gene Transcripts
- Gene Symbol
- CXXC1P1
- Entrez Gene ID
- 392459
- Gene Name
- CXXC finger protein 1 pseudogene 1
- Gene Aliases
- CXorf25, NCRNA00236
- Chromosome Location
- Chr. X: 47707191 - 47736628 on Build GRCh38
- UniGene ID
- Hs.562045
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_033924.1 |
|
2 |
342 |
|
GenBank mRNA |
AK094108.1 |
- |
2 |
344 |
|
View Full Product Details