Gene Transcripts

Gene Symbol
DTHD1
Entrez Gene ID
401124
Gene Name
death domain containing 1
Gene Aliases
Chromosome Location
Chr. 4: 36281593 - 36347511 on Build GRCh38
UniGene ID
Hs.363407
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136536.4 NP_001130008.2 1, 2 423
XM_011513696.2 XP_011511998.1 1, 2 441
GenBank mRNA AK131492.1 - 1, 2 416
AK304357.1 - 1, 2 409
AK316523.1 - 1, 2 409
DC417013.1 - 1, 2 423

View Full Product Details