Gene Transcripts
- Gene Symbol
- ANKRD26
- Entrez Gene ID
- 22852
- Gene Name
- ankyrin repeat domain 26
- Gene Aliases
- THC2, bA145E8.1
- Chromosome Location
- Chr. 10: 26938195 - 27100498 on Build GRCh38
- UniGene ID
- Hs.361041
- Species
- Homo sapiens
- Gene Symbol
- LINC00202-1
- Entrez Gene ID
- 387644
- Gene Name
- long intergenic non-protein coding RNA 202-1
- Gene Aliases
- C10orf51, LINC00202, NCRNA00202, bB27G4.1
- Chromosome Location
- Chr. 10: 26931206 - 26942001 on Build GRCh38
- UniGene ID
- Hs.529965
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026795.1 |
|
1 |
635 |
|
GenBank mRNA |
AK128027.1 |
- |
1 |
1325 |
|
View Full Product Details