Gene Transcripts

Gene Symbol
ACER3
Entrez Gene ID
55331
Gene Name
alkaline ceramidase 3
Gene Aliases
APHC, PHCA
Chromosome Location
Chr. 11: 76860873 - 77026797 on Build GRCh38
UniGene ID
Hs.23862
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001300953.1 NP_001287882.1 10 2715
NM_001300954.1 NP_001287883.1 11 2897
NM_001300955.1 NP_001287884.1 10 2773
NM_018367.6 NP_060837.3 11 2826
XM_011545151.2 XP_011543453.1 10 2690
XM_011545152.2 XP_011543454.1 11 2762
GenBank mRNA AF214454.1 - 11 2781
AF327353.1 - 11 2791

View Full Product Details