Gene Transcripts
- Gene Symbol
- ATP6V1E2
- Entrez Gene ID
- 90423
- Gene Name
- ATPase H+ transporting V1 subunit E2
- Gene Aliases
- ATP6E1, ATP6EL2, ATP6V1EL2, VMA4
- Chromosome Location
- Chr. 2: 46511835 - 46542557 on Build GRCh38
- UniGene ID
- Hs.659656
- Species
- Homo sapiens
- Gene Symbol
- LOC440862
- Entrez Gene ID
- 440862
- Gene Name
- uncharacterized LOC440862
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.437691
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK123751.1 |
- |
1 |
113 |
|
View Full Product Details