Gene Transcripts

Gene Symbol
IDI2-AS1
Entrez Gene ID
55853
Gene Name
IDI2 antisense RNA 1
Gene Aliases
C10orf110, HT009, IDI2-AS
Chromosome Location
Chr. 10: 1022637 - 1044201 on Build GRCh38
UniGene ID
Hs.728538
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024628.1 4 1011
NR_024629.1 3 848
NR_027708.1 5 1144
NR_027709.1 5 1228
GenBank mRNA AF220183.1 - 3 819

View Full Product Details