Gene Transcripts
- Gene Symbol
- STAG3L4
- Entrez Gene ID
- 64940
- Gene Name
- stromal antigen 3-like 4 (pseudogene)
- Gene Aliases
- STAG3L4P
- Chromosome Location
- Chr. 7: 67302638 - 67321526 on Build GRCh38
- UniGene ID
- Hs.632013
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_040585.1 |
|
6 |
1667 |
|
NR_040586.1 |
|
5 |
1591 |
|
GenBank mRNA |
AK023257.1 |
- |
5 |
1585 |
|
View Full Product Details