Gene Transcripts

Gene Symbol
PTPMT1
Entrez Gene ID
114971
Gene Name
protein tyrosine phosphatase, mitochondrial 1
Gene Aliases
DUSP23, MOSP, PLIP, PNAS-129
Chromosome Location
Chr. 11: 47565430 - 47573461 on Build GRCh38
UniGene ID
Hs.656205
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175732.2 NP_783859.1 3 521
GenBank mRNA BC014048.1 - 2 167
BC020242.1 - 3 350 4591438
DA476984.1 - 3 377
DR004030.1 - 3 379

View Full Product Details