Gene Transcripts

Gene Symbol
TCP11
Entrez Gene ID
6954
Gene Name
t-complex 11
Gene Aliases
D6S230E, FPPR
Chromosome Location
Chr. 6: 35118071 - 35141642 on Build GRCh38
UniGene ID
Hs.435371
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001093728.2 NP_001087197.1 6 921
NM_001261817.1 NP_001248746.1 6 906
NM_001261818.1 NP_001248747.1 5 783
NM_001261819.1 NP_001248748.1 5 768
NM_001261820.1 NP_001248749.1 7 1127
NM_001261821.1 NP_001248750.1 7 1112
NM_018679.5 NP_061149.1 6 1015
XM_005249339.2 XP_005249396.1 5 676
XM_011514830.1 XP_011513132.1 6 1153
XM_011514831.1 XP_011513133.1 4 589
XM_011514832.1 XP_011513134.1 3 453
GenBank mRNA AF260330.1 - 3 464
AF269223.1 - 5 699
AF536532.1 - 7 967
AF536533.1 - 6 954
AK057234.1 - 6 738
AK098590.1 - 4 510
AK301778.1 - 6 797
AK301975.1 - 5 704
AK302022.1 - 5 669
AK302375.1 - 6 846
AK302456.1 - 6 827
AK315078.1 - 6 948
AK316403.1 - 6 730
AY069943.1 - 5 723
BC033729.1 - 6 867 5167179
BC040003.1 - 7 969 5741923
BC048418.1 - 6 945 5298037
BC090050.1 - 7 973 30387768

View Full Product Details