Gene Transcripts
- Gene Symbol
- TATDN3
- Entrez Gene ID
- 128387
- Gene Name
- TatD DNase domain containing 3
- Gene Aliases
- Chromosome Location
- Chr. 1: 212791828 - 212816825 on Build GRCh38
- UniGene ID
- Hs.530538
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001042552.2 |
NP_001036017.1
|
2, 3 |
193 |
|
NM_001042553.2 |
NP_001036018.1
|
2, 3 |
193 |
|
NM_001146169.1 |
NP_001139641.1
|
2, 3 |
193 |
|
NM_001146170.1 |
NP_001139642.1
|
2, 3 |
193 |
|
NM_001146171.1 |
NP_001139643.1
|
2, 3 |
193 |
|
XM_011509202.2 |
XP_011507504.1
|
2, 3 |
188 |
|
XM_017000328.1 |
XP_016855817.1
|
2, 3 |
188 |
|
XM_017000329.1 |
XP_016855818.1
|
2, 3 |
188 |
|
XM_017000330.1 |
XP_016855819.1
|
2, 3 |
188 |
|
XM_017000331.1 |
XP_016855820.1
|
2, 3 |
188 |
|
GenBank mRNA |
AK293129.1 |
- |
2, 3 |
193 |
|
AK295335.1 |
- |
1, 2 |
118 |
|
AK304557.1 |
- |
2, 3 |
151 |
|
AL832248.1 |
- |
2, 3 |
135 |
|
BC048115.1 |
- |
2, 3 |
104 |
|
BC113638.1 |
- |
2, 3 |
137 |
8322690 |
BC117485.1 |
- |
2, 3 |
137 |
40126036 |
BC143956.1 |
- |
2, 3 |
137 |
9052474 |
BC143957.1 |
- |
2, 3 |
137 |
|
DB456872.1 |
- |
2, 3 |
171 |
|
View Full Product Details