Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001100915.1 | NP_001094385.1 | 4 | 685 | |
GenBank mRNA | AK097481.1 | - | 4 | 688 | |
AK302748.1 | - | 4 | 669 | ||
BC070103.1 | - | 4 | 653 | ||
BC117335.1 | - | 4 | 685 | 40125886 |