Gene Transcripts

Gene Symbol
MPEG1
Entrez Gene ID
219972
Gene Name
macrophage expressed 1
Gene Aliases
MPG1, MPS1, Mpg-1
Chromosome Location
Chr. 11: 59208510 - 59213021 on Build GRCh38
UniGene ID
Hs.744920
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039396.1 NP_001034485.1 1 952
GenBank mRNA AK074166.1 - 1 882
AK095457.1 - 1 31
AK097881.1 - 1 849
BC104997.1 - 1 817 8144000
BC112230.1 - 1 817 8327698

View Full Product Details