Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001039396.1 | NP_001034485.1 | 1 | 1615 | |
GenBank mRNA | AK074166.1 | - | 1 | 1545 | |
AK095457.1 | - | 1 | 694 | ||
AK097881.1 | - | 1 | 1512 | ||
BC104997.1 | - | 1 | 1480 | 8144000 | |
BC112230.1 | - | 1 | 1480 | 8327698 |