Gene Transcripts

Gene Symbol
OTUD1
Entrez Gene ID
220213
Gene Name
OTU deubiquitinase 1
Gene Aliases
DUBA7, OTDC1
Chromosome Location
Chr. 10: 23439269 - 23442381 on Build GRCh38
UniGene ID
Hs.499042
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145373.2 NP_001138845.1 1 1550
GenBank mRNA AB188491.1 - 1 1550
AK096389.1 - 1 395

View Full Product Details