Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001135924.1 | NP_001129396.1 | 5 | 1106 | |
XM_005249641.1 | XP_005249698.1 | 5 | 783 | ||
XM_011515174.1 | XP_011513476.1 | 5 | 783 | ||
XM_011515175.1 | XP_011513477.1 | 5 | 783 | ||
XM_017011828.1 | XP_016867317.1 | 5 | 783 | ||
XM_017011829.1 | XP_016867318.1 | 4 | 452 | ||
XM_017011830.1 | XP_016867319.1 | 5 | 783 | ||
GenBank mRNA | AK075525.1 | - | 7 | 823 | |
BC144316.1 | - | 5 | 821 | ||
BC144319.1 | - | 5 | 821 | 9052839 |