Gene Transcripts

Gene Symbol
OBSL1
Entrez Gene ID
23363
Gene Name
obscurin like 1
Gene Aliases
Chromosome Location
Chr. 2: 219549408 - 219571573 on Build GRCh38
UniGene ID
Hs.526594
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017003696.1 XP_016859185.1 21 6250
GenBank mRNA AB014557.2 - 16 5052
F30329.1 - 1 29

View Full Product Details