Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018686.5 | NP_061156.1 | 6 | 1021 | |
GenBank mRNA | AF271388.1 | - | 6 | 909 | |
AF397212.1 | - | 6 | 891 | ||
AK022927.1 | - | 6 | 931 | ||
AK222981.1 | - | 6 | 920 | ||
AL832975.1 | - | 6 | 969 |