Gene Transcripts
- Gene Symbol
- CD58
- Entrez Gene ID
- 965
- Gene Name
- CD58 molecule
- Gene Aliases
- LFA-3, LFA3, ag3
- Chromosome Location
- Chr. 1: 116487140 - 116571093 on Build GRCh38
- UniGene ID
- Hs.34341
- Species
- Homo sapiens
- Gene Symbol
- LOC100506459
- Entrez Gene ID
- 100506459
- Gene Name
- uncharacterized LOC100506459
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.734297
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK126833.1 |
- |
2 |
1940 |
|
AL832882.1 |
- |
1 |
4032 |
|
- Gene Symbol
- LOC105378922
- Entrez Gene ID
- 105378922
- Gene Name
- uncharacterized LOC105378922
- Gene Aliases
- Chromosome Location
- Chr. 1: 116493016 - 116505088 on Build GRCh38
- UniGene ID
- Hs.734297
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC018791.1 |
- |
1 |
879 |
|
View Full Product Details