Gene Transcripts

Gene Symbol
H2BFM
Entrez Gene ID
286436
Gene Name
H2B histone family member M
Gene Aliases
Chromosome Location
Chr. X: 104039753 - 104042454 on Build GRCh38
UniGene ID
Hs.376474
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164416.1 NP_001157888.1 3 1681
XM_006724640.2 XP_006724703.1 4 709
XM_011530922.2 XP_011529224.1 3 853
GenBank mRNA AK093522.1 - 3 1681

View Full Product Details