Gene Transcripts

Gene Symbol
FMN1
Entrez Gene ID
342184
Gene Name
formin 1
Gene Aliases
FMN, LD
Chromosome Location
Chr. 15: 32765544 - 33194765 on Build GRCh38
UniGene ID
Hs.657649
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AL833157.1 - 2 1776
BC101971.1 - 2 362
BC103692.1 - 2 1645
BC107593.1 - 2 1645

View Full Product Details