Gene Transcripts

Gene Symbol
DCDC2C
Entrez Gene ID
728597
Gene Name
doublecortin domain containing 2C
Gene Aliases
Chromosome Location
Chr. 2: 3703592 - 3847411 on Build GRCh38
UniGene ID
Hs.304233
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001287444.1 NP_001274373.1 11 1343
GenBank mRNA BG202560.1 - 1 104

View Full Product Details