Gene Transcripts

Gene Symbol
IL17RB
Entrez Gene ID
55540
Gene Name
interleukin 17 receptor B
Gene Aliases
CRL4, EVI27, IL17BR, IL17RH1
Chromosome Location
Chr. 3: 53846494 - 53866526 on Build GRCh38
UniGene ID
Hs.654970
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018725.3 NP_061195.2 4 349
XM_005265310.4 XP_005265367.1 4 349
XM_005265311.4 XP_005265368.1 4 374
XM_005265312.4 XP_005265369.1 4 405
XM_011533941.2 XP_011532243.1 4 349
XM_017006807.1 XP_016862296.1 4 349
GenBank mRNA AF208110.1 - 4 315
AF208111.1 - 4 315
AF212365.1 - 4 319
AF250309.1 - 4 310
AK307749.1 - 4 331
AY359122.1 - 4 316
BC000980.1 - 4 319 2985728
DA093617.1 - 4 349

View Full Product Details