Gene Transcripts
- Gene Symbol
- ABCA17P
- Entrez Gene ID
- 650655
- Gene Name
- ATP binding cassette subfamily A member 17, pseudogene
- Gene Aliases
- ABCA17
- Chromosome Location
- Chr. 16: 2340922 - 2426699 on Build GRCh38
- UniGene ID
- Hs.376281
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003574.1 |
|
5 |
918 |
|
GenBank mRNA |
DQ266102.1 |
- |
5 |
918 |
|
View Full Product Details