Gene Transcripts

Gene Symbol
TTC34
Entrez Gene ID
100287898
Gene Name
tetratricopeptide repeat domain 34
Gene Aliases
Chromosome Location
Chr. 1: 2636988 - 2789737 on Build GRCh38
UniGene ID
Hs.632363
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242672.1 NP_001229601.1 7 1614
XM_016999990.1 XP_016855479.1 7 1686

View Full Product Details