Gene Transcripts

Gene Symbol
RP1
Entrez Gene ID
6101
Gene Name
retinitis pigmentosa 1 (autosomal dominant)
Gene Aliases
DCDC4A, ORP1
Chromosome Location
Chr. 8: 54509448 - 54630834 on Build GRCh38
UniGene ID
Hs.732820
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK098176.1 - 8 1093
AK302348.1 - 5 693
AK304443.1 - 7 854
AK316511.1 - 7 1288

View Full Product Details