Gene Transcripts

Gene Symbol
DTHD1
Entrez Gene ID
401124
Gene Name
death domain containing 1
Gene Aliases
Chromosome Location
Chr. 4: 36281593 - 36347511 on Build GRCh38
UniGene ID
Hs.363407
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136536.4 NP_001130008.2 2 697
NM_001170700.2 NP_001164171.1 2 852
XM_006714014.3 XP_006714077.1 3 1331
XM_011513693.2 XP_011511995.1 3 1332
XM_011513694.2 XP_011511996.1 3 1332
XM_011513695.2 XP_011511997.1 3 1331
XM_011513696.2 XP_011511998.1 2 715
XM_017008191.1 XP_016863680.1 3 1332
XM_017008192.1 XP_016863681.1 3 1334
XM_017008193.1 XP_016863682.1 3 1333
GenBank mRNA AK131492.1 - 2 690
AK304357.1 - 2 683
AK316523.1 - 2 683
BC157885.1 - 2 852 9057096

View Full Product Details