Gene Transcripts

Gene Symbol
AACSP1
Entrez Gene ID
729522
Gene Name
acetoacetyl-CoA synthetase pseudogene 1
Gene Aliases
AACSL
Chromosome Location
Chr. 5: 178764861 - 178818435 on Build GRCh38
UniGene ID
Hs.382790
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024035.2 9 1291
NR_135095.1 9 1294
GenBank mRNA BC140764.1 - 9 1245
BC144474.1 - 9 1244

View Full Product Details