Gene Transcripts

Gene Symbol
SNTG1
Entrez Gene ID
54212
Gene Name
syntrophin gamma 1
Gene Aliases
G1SYN, SYN4
Chromosome Location
Chr. 8: 49909726 - 50796656 on Build GRCh38
UniGene ID
Hs.584914
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001287813.2 NP_001274742.1 6 641
NM_001287814.2 NP_001274743.1 4 793
NM_001321773.1 NP_001308702.1 4 793
NM_001321775.1 NP_001308704.1 6 641
NM_001321776.1 NP_001308705.1 5 868
NM_001321777.1 NP_001308706.1 4 793
NM_001321778.1 NP_001308707.1 5 599
NM_018967.4 NP_061840.1 5 868
NR_135794.1 5 848
NR_135795.1 5 868
NR_135796.1 5 868
NR_135797.1 5 848
NR_135798.1 4 793
XM_011517548.2 XP_011515850.1 4 978
XM_017013579.1 XP_016869068.1 7 3797
XM_017013580.1 XP_016869069.1 5 391
XM_017013581.1 XP_016869070.1 5 1113
XM_017013582.1 XP_016869071.1 5 1113
GenBank mRNA AJ003030.1 - 4 429
AK314084.1 - 6 578
AL161971.1 - 5 763
BC075072.2 - 3 233 30915592
BC104829.1 - 4 315 8143832
BC143695.1 - 4 315

View Full Product Details