Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001256165.1 | NP_001243094.1 | 2 | 474 | |
NM_019026.4 | NP_061899.2 | 2 | 274 | ||
NR_045818.1 | 2 | 197 | |||
GenBank mRNA | AB020980.1 | - | 2 | 241 | |
AF070626.1 | - | 2 | 230 | ||
AF274935.1 | - | 2 | 110 | ||
AF277194.1 | - | 2 | 148 | ||
AK298614.1 | - | 2 | 441 | ||
AK316080.1 | - | 2 | 407 | ||
AK316610.1 | - | 2 | 220 | ||
AY359027.1 | - | 2 | 209 | ||
AY763589.1 | - | 2 | 203 | ||
BC000104.2 | - | 2 | 194 | 3504808 | |
BG772940.1 | - | 2 | 279 | ||
BP257387.1 | - | 2 | 170 | ||
CN430121.1 | - | 2 | 252 |