Gene Transcripts
- Gene Symbol
- FLJ43315
- Entrez Gene ID
- 644316
- Gene Name
- asparagine synthetase pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 9: 64016950 - 64061612 on Build GRCh38
- UniGene ID
- Hs.632789
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC057848.1 |
- |
4 |
685 |
|
View Full Product Details