Gene Transcripts

Gene Symbol
IQCF6
Entrez Gene ID
440956
Gene Name
IQ motif containing F6
Gene Aliases
Chromosome Location
Chr. 3: 51778561 - 51779791 on Build GRCh38
UniGene ID
Hs.444316
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001143833.3 NP_001137305.2 1 436
GenBank mRNA BX093815.1 - 1 436

View Full Product Details