Gene Transcripts
- Gene Symbol
- C5orf42
- Entrez Gene ID
- 65250
- Gene Name
- chromosome 5 open reading frame 42
- Gene Aliases
- Hug, JBTS17, OFD6
- Chromosome Location
- Chr. 5: 37063928 - 37249428 on Build GRCh38
- UniGene ID
- Hs.586199
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_023073.3 |
NP_075561.3
|
11, 12 |
1749 |
|
XM_005248345.3 |
XP_005248402.1
|
11, 12 |
1737 |
|
XM_005248346.3 |
XP_005248403.1
|
11, 12 |
1657 |
|
XM_005248347.3 |
XP_005248404.1
|
11, 12 |
1657 |
|
XM_005248349.3 |
XP_005248406.1
|
11, 12 |
1657 |
|
XM_005248350.3 |
XP_005248407.1
|
11, 12 |
1655 |
|
XM_011514085.2 |
XP_011512387.1
|
11, 12 |
1656 |
|
XM_011514086.2 |
XP_011512388.1
|
11, 12 |
1653 |
|
XM_011514087.2 |
XP_011512389.1
|
11, 12 |
1742 |
|
XM_011514088.2 |
XP_011512390.1
|
11, 12 |
1742 |
|
XM_011514089.2 |
XP_011512391.1
|
11, 12 |
1742 |
|
XM_011514090.2 |
XP_011512392.1
|
11, 12 |
1520 |
|
XM_011514092.2 |
XP_011512394.1
|
11, 12 |
1742 |
|
XM_017009760.1 |
XP_016865249.1
|
10, 11 |
1468 |
|
XM_017009761.1 |
XP_016865250.1
|
10, 11 |
1436 |
|
XM_017009762.1 |
XP_016865251.1
|
7, 8 |
1195 |
|
XM_017009763.1 |
XP_016865252.1
|
7, 8 |
1068 |
|
XM_017009764.1 |
XP_016865253.1
|
7, 8 |
1029 |
|
XM_017009765.1 |
XP_016865254.1
|
11, 12 |
1437 |
|
GenBank mRNA |
DN831769.1 |
- |
1, 2 |
95 |
|
View Full Product Details