Gene Transcripts
- Gene Symbol
- PUS7
- Entrez Gene ID
- 54517
- Gene Name
- pseudouridylate synthase 7 (putative)
- Gene Aliases
- Chromosome Location
- Chr. 7: 105456501 - 105522270 on Build GRCh38
- UniGene ID
- Hs.520619
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001318163.1 |
NP_001305092.1
|
12, 13 |
1556 |
|
NM_001318164.1 |
NP_001305093.1
|
12, 13 |
1745 |
|
NM_019042.4 |
NP_061915.2
|
12, 13 |
1769 |
|
XM_005250462.3 |
XP_005250519.1
|
13, 14 |
1795 |
|
XM_017012367.1 |
XP_016867856.1
|
13, 14 |
1723 |
|
GenBank mRNA |
AB067484.1 |
- |
12, 13 |
1483 |
|
AK000492.1 |
- |
2, 3 |
273 |
|
AK091283.1 |
- |
12, 13 |
1749 |
|
AK128629.1 |
- |
12, 13 |
1731 |
|
BC005209.2 |
- |
4, 5 |
482 |
|
BC011396.1 |
- |
12, 13 |
1725 |
3870711 |
View Full Product Details