Gene Transcripts
- Gene Symbol
- TRMT13
- Entrez Gene ID
- 54482
- Gene Name
- tRNA methyltransferase 13 homolog
- Gene Aliases
- CCDC76
- Chromosome Location
- Chr. 1: 100133080 - 100150498 on Build GRCh38
- UniGene ID
- Hs.729095
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_019083.2 |
NP_061956.2
|
1, 2 |
174 |
|
NR_135078.1 |
|
1, 2 |
174 |
|
XM_005270945.3 |
XP_005271002.1
|
1, 2 |
244 |
|
XM_005270946.2 |
XP_005271003.1
|
1, 2 |
164 |
|
XM_017001504.1 |
XP_016856993.1
|
1, 2 |
161 |
|
XM_017001505.1 |
XP_016856994.1
|
1, 2 |
192 |
|
GenBank mRNA |
AK001149.1 |
- |
1, 2 |
159 |
|
AK002081.1 |
- |
1, 2 |
161 |
|
AK289404.1 |
- |
1, 2 |
161 |
|
AK294738.1 |
- |
1, 2 |
159 |
|
AK295828.1 |
- |
1, 2 |
171 |
|
AK298941.1 |
- |
1, 2 |
159 |
|
AL699548.1 |
- |
1, 2 |
175 |
|
BC057226.1 |
- |
1, 2 |
174 |
|
BC075811.1 |
- |
1, 2 |
171 |
5104751 |
View Full Product Details