Gene Transcripts
- Gene Symbol
- ZKSCAN4
- Entrez Gene ID
- 387032
- Gene Name
- zinc finger with KRAB and SCAN domains 4
- Gene Aliases
- P1P373C6, ZNF307, ZNF427, ZSCAN36
- Chromosome Location
- Chr. 6: 28241697 - 28259312 on Build GRCh38
- UniGene ID
- Hs.44720
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001304506.1 |
NP_001291435.1
|
3, 4 |
313 |
|
NM_019110.4 |
NP_061983.2
|
3, 4 |
978 |
|
XM_005249095.3 |
XP_005249152.1
|
5, 6 |
1022 |
|
XM_005249097.3 |
XP_005249154.1
|
5, 6 |
718 |
|
XM_005249098.3 |
XP_005249155.1
|
3, 4 |
636 |
|
XM_005249100.4 |
XP_005249157.2
|
3, 4 |
369 |
|
XM_006715095.3 |
XP_006715158.1
|
4, 5 |
495 |
|
XM_011514588.2 |
XP_011512890.1
|
3, 4 |
878 |
|
XM_011514589.2 |
XP_011512891.1
|
3, 4 |
434 |
|
XM_017010844.1 |
XP_016866333.1
|
5, 6 |
1673 |
|
GenBank mRNA |
AK302017.1 |
- |
3, 4 |
294 |
|
AK315783.1 |
- |
3, 4 |
948 |
|
AY781778.1 |
- |
3, 4 |
948 |
|
BC014031.2 |
- |
3, 4 |
906 |
4336904 |
KU178905.1 |
- |
3, 4 |
662 |
|
View Full Product Details