Gene Transcripts
- Gene Symbol
- ATXN7L3
- Entrez Gene ID
- 56970
- Gene Name
- ataxin 7 like 3
- Gene Aliases
- Chromosome Location
- Chr. 17: 44191805 - 44199976 on Build GRCh38
- UniGene ID
- Hs.512651
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001098833.1 |
NP_001092303.1
|
4, 5 |
553 |
|
NM_020218.1 |
NP_064603.1
|
4, 5 |
553 |
|
XM_011525038.2 |
XP_011523340.1
|
4, 5 |
1147 |
|
XM_011525044.2 |
XP_011523346.1
|
4, 5 |
1147 |
|
XM_011525046.2 |
XP_011523348.1
|
4, 5 |
1147 |
|
XM_017024880.1 |
XP_016880369.1
|
5, 6 |
725 |
|
XM_017024881.1 |
XP_016880370.1
|
5, 6 |
1024 |
|
XM_017024882.1 |
XP_016880371.1
|
4, 5 |
1147 |
|
XM_017024883.1 |
XP_016880372.1
|
5, 6 |
728 |
|
XM_017024884.1 |
XP_016880373.1
|
4, 5 |
771 |
|
XM_017024885.1 |
XP_016880374.1
|
5, 6 |
728 |
|
XM_017024886.1 |
XP_016880375.1
|
5, 6 |
728 |
|
GenBank mRNA |
AK056002.1 |
- |
1, 2 |
97 |
|
AK126117.1 |
- |
4, 5 |
771 |
|
BC037418.1 |
- |
4, 5 |
460 |
|
BC113595.1 |
- |
4, 5 |
553 |
8322647 |
BC126113.1 |
- |
4, 5 |
553 |
8991829 |
View Full Product Details