Gene Transcripts

Gene Symbol
PLEKHA3
Entrez Gene ID
65977
Gene Name
pleckstrin homology domain containing A3
Gene Aliases
FAPP1
Chromosome Location
Chr. 2: 178480410 - 178516324 on Build GRCh38
UniGene ID
Hs.41086
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019091.3 NP_061964.3 8 2401
XM_011511689.2 XP_011509991.1 9 2510
GenBank mRNA AF286162.1 - 8 2238
AK000074.1 - 6 1804
DQ786259.1 - 5 1696
DQ786318.1 - 4 1558

View Full Product Details