Gene Transcripts
- Gene Symbol
- SEPT5-GP1BB
- Entrez Gene ID
- 100526833
- Gene Name
- SEPT5-GP1BB readthrough
- Gene Aliases
- Chromosome Location
- Chr. 22: 19717220 - 19724774 on Build GRCh38
- UniGene ID
- Hs.283743
- Species
- Homo sapiens
- Gene Symbol
- GP1BB
- Entrez Gene ID
- 2812
- Gene Name
- glycoprotein Ib platelet beta subunit
- Gene Aliases
- BDPLT1, BS, CD42C, GPIBB, GPIbbeta
- Chromosome Location
- Chr. 22: 19723543 - 19724774 on Build GRCh38
- UniGene ID
- Hs.283743
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000407.4 |
NP_000398.1
|
2 |
931 |
|
GenBank mRNA |
J03259.1 |
- |
2 |
926 |
|
View Full Product Details