Gene Transcripts

Gene Symbol
C9orf85
Entrez Gene ID
138241
Gene Name
chromosome 9 open reading frame 85
Gene Aliases
Chromosome Location
Chr. 9: 71911340 - 71982700 on Build GRCh38
UniGene ID
Hs.534190
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182505.3 NP_872311.2 4 1107
XM_011518226.2 XP_011516528.1 5 1405

View Full Product Details