Gene Transcripts

Gene Symbol
CDRT1
Entrez Gene ID
374286
Gene Name
CMT1A duplicated region transcript 1
Gene Aliases
C170RF1, C17ORF1, C17ORF1A, FBXW10B, FBXW10P1, HREP, SM25H2
Chromosome Location
Chr. 17: 15565482 - 15619704 on Build GRCh38
UniGene ID
Hs.548021
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006382.3 NP_006373.2 11, 12 2198
GenBank mRNA BC143291.1 - 4, 5 474
U43383.1 - 4, 5 492
U65652.1 - 4, 5 727

View Full Product Details