Gene Transcripts

Gene Symbol
CBWD3
Entrez Gene ID
445571
Gene Name
COBW domain containing 3
Gene Aliases
bA561O23.1
Chromosome Location
Chr. 9: 68232437 - 68300035 on Build GRCh38
UniGene ID
Hs.645337
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_005277639.2 XP_005277696.1 7, 8 734
XM_005277640.2 XP_005277697.1 7, 8 734
XM_017014730.1 XP_016870219.1 6, 7 674
XM_017014732.1 XP_016870221.1 2, 3 185

Gene Symbol
CBWD1
Entrez Gene ID
55871
Gene Name
COBW domain containing 1
Gene Aliases
COBP
Chromosome Location
Chr. 9: 121038 - 179083 on Build GRCh38
UniGene ID
Hs.531330
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_005251514.1 XP_005251571.1 7, 8 674
XM_005251515.1 XP_005251572.1 7, 8 674
XM_011517964.2 XP_011516266.1 7, 8 674

Gene Symbol
CBWD2
Entrez Gene ID
150472
Gene Name
COBW domain containing 2
Gene Aliases
Chromosome Location
Chr. 2: 113437691 - 113496204 on Build GRCh38
UniGene ID
Hs.531330
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_005263602.2 XP_005263659.1 7, 8 746
XM_005263604.2 XP_005263661.1 7, 8 746

Gene Symbol
CBWD5
Entrez Gene ID
220869
Gene Name
COBW domain containing 5
Gene Aliases
DC36
Chromosome Location
Chr. 9: 65666374 - 65734041 on Build GRCh38
UniGene ID
Hs.355950
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024916.3 NP_001020087.2 7, 8 711
XM_005272751.1 XP_005272808.1 7, 8 671
XM_017014455.1 XP_016869944.1 7, 8 671
XM_017014457.1 XP_016869946.1 8, 9 602
XM_017014460.1 XP_016869949.1 7, 8 671
XM_017014461.1 XP_016869950.1 9, 10 850
GenBank mRNA BC082271.1 - 7, 8 607

Gene Symbol
CBWD6
Entrez Gene ID
644019
Gene Name
COBW domain containing 6
Gene Aliases
Chromosome Location
Chr. 9: 41131309 - 41198865 on Build GRCh38
UniGene ID
Hs.597638
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017015022.1 XP_016870511.1 6, 7 674
XM_017015024.1 XP_016870513.1 6, 7 674
XM_017015025.1 XP_016870514.1 6, 7 674
XM_017015033.1 XP_016870522.1 2, 3 185
XM_017015036.1 XP_016870525.1 7, 8 734
GenBank mRNA AK293173.1 - 7, 8 669
BC069006.1 - 8, 9 779

View Full Product Details