Gene Transcripts

Gene Symbol
SFTA2
Entrez Gene ID
389376
Gene Name
surfactant associated 2
Gene Aliases
GSGL541, SFTPG, SP-G, UNQ541
Chromosome Location
Chr. 6: 30931350 - 30932175 on Build GRCh38
UniGene ID
Hs.211267
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_205854.2 NP_995326.1 2 174
GenBank mRNA AY102070.1 - 2 149
AY359057.1 - 2 152
BC039677.1 - 2 174 5186107

View Full Product Details