Gene Transcripts

Gene Symbol
C21orf62
Entrez Gene ID
56245
Gene Name
chromosome 21 open reading frame 62
Gene Aliases
B37, C21orf120, PRED81
Chromosome Location
Chr. 21: 32790673 - 32813808 on Build GRCh38
UniGene ID
Hs.517235
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001162495.2 NP_001155967.2 4 456
NM_001162496.2 NP_001155968.2 2 300
NM_019596.5 NP_062542.5 3 381
GenBank mRNA AF231922.1 - 3 287
AK290983.1 - 3 324
CN428551.1 - 3 327
DQ656042.2 - 3 381

View Full Product Details